2-Fluoro-6-hydroxybenzonitrile 98,0 GC, T

Supplier: TCI America
Danger

F0985-1G F0985-5G
TCF0985-1GEA 34.76 CAD
TCF0985-1G TCF0985-5G
2-Fluoro-6-hydroxybenzonitrile 98,0 GC, T
2-Fluoro-6-hydroxybenzonitrile
Formula: C₇H₄FNO
MW: 137.11 g/mol
MDL Number: MFCD03428592
CAS Number: 140675-43-0
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 155.0 to 159.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR