4-Ethoxy-3-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

E1124-1G E1124-5G
TCE1124-1GEA 37.92 CAD
TCE1124-1G TCE1124-5G
4-Ethoxy-3-fluorophenylboronic acid (contains varying amounts of Anhydride)
4-Ethoxy-3-fluorophenylboronic acid
Formula: C₈H₁₀BFO₃
MW: 183.97
MDL Number: MFCD04115667
CAS Number: 279263-10-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR