4-Bromothiazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B4594-1G B4594-5G
TCB4594-1GEA 67.94 CAD
TCB4594-1G TCB4594-5G
4-Bromothiazole ≥98.0% (by GC)
4-Bromothiazole
Formula: C₃H₂BrNS
Boiling Pt: 190°C
Flash Pt: 101°C
MDL Number: MFCD06657592
CAS Number: 34259-99-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR