5-Ethoxy-2-fluorophenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

E1235-1G E1235-5G
TCE1235-1GEA 102.7 CAD
TCE1235-1G TCE1235-5G
5-Ethoxy-2-fluorophenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)
5-Ethoxy-2-fluorophenylboronic acid
Formula: C₈H₁₀BFO₃
MDL Number: MFCD05664321
CAS Number: 900174-60-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 98.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR