3-Vinylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

V0141-1G V0141-5G
TCV0141-1GEA 107.44 CAD
TCV0141-1G TCV0141-5G
3-Vinylphenylboronic acid (contains varying amounts of Anhydride)
3-Vinylphenylboronic acid
Formula: C₈H₉BO₂
Melting Pt: 145°C
MDL Number: MFCD01074679
CAS Number: 15016-43-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 96.0 to 114.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR