3-Chloro-2-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

C2946-1G C2946-5G
TCC2946-1GEA 66.36 CAD
TCC2946-1G TCC2946-5G
3-Chloro-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Chloro-2-fluorophenylboronic acid
Formula: C₆H₅BClFO₂
Melting Pt: 247°C
MDL Number: MFCD05664224
CAS Number: 352535-82-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR