2-Ethoxy-4-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

E1204-1G E1204-5G
TCE1204-1GEA 148.52 CAD
TCE1204-1G TCE1204-5G
2-Ethoxy-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
2-Ethoxy-4-fluorophenylboronic acid
Formula: C₈H₁₀BFO₃
MDL Number: MFCD03095001
CAS Number: 480438-58-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 96.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR