2-Vinylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

V0140-1G V0140-5G
TCV0140-1GEA 167.48 CAD
TCV0140-1G TCV0140-5G
2-Vinylphenylboronic acid (contains varying amounts of Anhydride)
2-Vinylphenylboronic acid
Formula: C₈H₉BO₂
Melting Pt: 109°C
MDL Number: MFCD01074659
CAS Number: 15016-42-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 114.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR