2-Chloro-6-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

C3247-1G C3247-5G
TCC3247-1GEA 85.32 CAD
TCC3247-1G TCC3247-5G
2-Chloro-6-fluorophenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-6-fluorophenylboronic acid
Formula: C₆H₅BClFO₂
MDL Number: MFCD04039892
CAS Number: 313545-32-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR