4-Chloro-2-methylbenzyl alcohol ≥98.0% (by GC)

Supplier: TCI America
Warning

C3069-1G C3069-5G
TCC3069-1GEA 112.18 CAD
TCC3069-1G TCC3069-5G
4-Chloro-2-methylbenzyl alcohol ≥98.0% (by GC)
4-Chloro-2-methylbenzyl alcohol
Formula: C₈H₉ClO
MDL Number: MFCD06201140
CAS Number: 129716-11-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR