1-Butyl-3-methylimidazolium dicyanamide ≥96.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

B5447-25G B5447-5G
76219-610EA 594.08 CAD
76219-610 76219-612
1-Butyl-3-methylimidazolium dicyanamide ≥96.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazolium dicyanamide
Formula: C₁₀H₁₅N₅
Melting Pt: -6°C
MDL Number: MFCD07784457
CAS Number: 448245-52-1

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (HPLC) min. 96.0 % (total of Ion-Pair)
Purity (with Total Nitrogen) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR