3-Amino-4-methylbenzyl alcohol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-Amino-4-methylbenzyl Alcohol

A2998-1G A2998-5G
TCA2998-1GEA 75.84 CAD
TCA2998-1G TCA2998-5G
3-Amino-4-methylbenzyl alcohol ≥98.0% (by GC, titration analysis)
3-Amino-4-methylbenzyl alcohol
Formula: C₈H₁₁NO
MDL Number: MFCD00075051
CAS Number: 81863-45-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR