4,5-Dimethoxy-2-nitrobenzyl alcohol ≥98.0% (by GC)
Supplier: TCI America
D4811-1G
D4811-5G
TCD4811-1GEA
94.8
CAD
TCD4811-1G
TCD4811-5G
4,5-Dimethoxy-2-nitrobenzyl alcohol ≥98.0% (by GC)
4,5-Dimethoxy-2-nitrobenzyl alcohol
Formula:
C₉H₁₁NO₅ |
MDL Number:
MFCD00014701 CAS Number: 1016-58-6 EINECS: 213-806-4 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...