5-Amino-3-methylisoxazole ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A2867-1G A2867-5G
TCA2867-1GEA 94.8 CAD
TCA2867-1G TCA2867-5G
5-Amino-3-methylisoxazole ≥97.0% (by titrimetric analysis)
5-Amino-3-methylisoxazole
Formula: C₄H₆N₂O
MDL Number: MFCD00003151
CAS Number: 14678-02-5
EINECS: 238-719-9

Order Now

Specification Test Results

Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR