3-Chloropyridine-2-carbonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

C3022-1G C3022-5G
TCC3022-1GEA 26.86 CAD
TCC3022-1G TCC3022-5G
3-Chloropyridine-2-carbonitrile ≥98.0% (by GC)
3-Chloropyridine-2-carbonitrile
Formula: C₆H₃ClN₂
Boiling Pt: 100°C
MDL Number: MFCD03788834
CAS Number: 38180-46-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 81.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR