3-Bromo-2-thenoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4605-1G B4605-5G
TCB4605-1GEA 37.92 CAD
TCB4605-1G TCB4605-5G
3-Bromo-2-thenoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-2-thenoic acid
Formula: C₅H₃BrO₂S
MDL Number: MFCD00052291
CAS Number: 7311-64-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 196.0 to 200.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR