6-Fluoro-o-anisaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
F1175-1G
F1175-5G
TCF1175-1GEA
55.3
CAD
TCF1175-1G
TCF1175-5G
6-Fluoro-o-anisaldehyde ≥98.0% (by GC)
6-Fluoro-o-anisaldehyde
Formula:
C₈H₇FO₂ |
MDL Number:
MFCD01090998 CAS Number: 146137-74-8 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 59.0 to 63.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...