1-Ethyl-3-methylimidazolium methylsulfate ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
E1308-25G
E1308-5G
76220-058EA
178.54
CAD
76220-058
76220-060
1-Ethyl-3-methylimidazolium methylsulfate ≥98.0% (by HPLC)
1-Ethyl-3-methylimidazolium methyl sulphate
Formula:
C₇H₁₄N₂O₄S |
MDL Number:
MFCD08457697 CAS Number: 516474-01-4 |
Specification Test Results
Appearance | White to Almost white clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...