1-Decyl-3-methylimidazolium chloride ≥96.0% (by HPLC)

Supplier: TCI America
Warning

D5351-25G D5351-5G
76219-940EA 252.8 CAD
76219-940 76219-942
1-Decyl-3-methylimidazolium chloride ≥96.0% (by HPLC)
1-Decyl-3-methylimidazolium chloride
Formula: C₁₄H₂₇ClN₂
MDL Number: MFCD03427605
CAS Number: 171058-18-7

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR