4-Fluoro-o-toluonitrile ≥99.0% (by GC)
Supplier: TCI America
|
Danger
|
F0962-1G
F0962-5G
TCF0962-1GEA
47.4
CAD
TCF0962-1G
TCF0962-5G
4-Fluoro-o-toluonitrile ≥99.0% (by GC)
4-Fluoro-o-toluonitrile
Formula:
C₈H₆FN |
MDL Number:
MFCD03095106 CAS Number: 147754-12-9 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...