3-Chlorobenzoylacetonitrile ≥97.0% (by GC)

Supplier: TCI America
Warning

C2581-1G C2581-5G
TCC2581-1GEA 72.68 CAD
TCC2581-1G TCC2581-5G
3-Chlorobenzoylacetonitrile ≥97.0% (by GC)
3-Chlorobenzoylacetonitrile
Formula: C₉H₆ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD00067891
CAS Number: 21667-62-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 80.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR