2-(3-Chlorophenyl)ethanol ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Chlorophenethyl alcohol

C2907-25G C2907-5G
TCC2907-25GEA 487.59 CAD
TCC2907-25G TCC2907-5G
2-(3-Chlorophenyl)ethanol ≥98.0% (by GC)
2-(3-Chlorophenyl)ethanol
Formula: C₈H₉ClO
MW: 156.61
Boiling Pt: 257
Density: 1.189
MDL Number: MFCD00002892
CAS Number: 5182-44-5
EINECS: 225-964-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR