4-Chloro-3-fluorobenzotrifluoride ≥98.0% (by GC)

Supplier: TCI America
Warning

C3216-1G C3216-5G
TCC3216-1GEA 79.32 CAD
TCC3216-1G TCC3216-5G
4-Chloro-3-fluorobenzotrifluoride ≥98.0% (by GC)
4-Chloro-3-fluorobenzotrifluoride
Formula: C₇H₃ClF₄
MW: 198.55
MDL Number: MFCD00060646
CAS Number: 32137-20-5
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR