4'-Bromovalerophenone ≥98.0% (by GC)
Supplier: TCI America
B4733-25G
B4733-5G
TCB4733-25GEA
127.35
CAD
TCB4733-25G
TCB4733-5G
4'-Bromovalerophenone ≥98.0% (by GC)
4'-Bromovalerophenone
Formula:
C₁₁H₁₃BrO Boiling Pt: 170°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000107 CAS Number: 7295-44-5 EINECS: 230-729-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 36.0 to 40.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...