5-Bromo-o-toluonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

B4808-1G B4808-5G
TCB4808-1GEA 61.62 CAD
TCB4808-1G TCB4808-5G
5-Bromo-o-toluonitrile ≥98.0% (by GC)
5-Bromo-o-toluonitrile
Formula: C₈H₆BrN
MW: 196.05
MDL Number: MFCD08061920
CAS Number: 156001-51-3
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR