5-Methoxy-o-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Amino-4-methoxytoluene, 3-Amino-4-methylanisole, 1-Amino-3-methoxy-6-methylbenzene, 6-Methyl-m-anisidine, 5-Methoxy-2-methylaniline

M3009-1G M3009-5G
TCM3009-1GEA 91.64 CAD
TCM3009-1G TCM3009-5G
5-Methoxy-o-toluidine ≥98.0% (by GC, titration analysis)
5-Methoxy-o-toluidine
Formula: C₈H₁₁NO
MW: 137.18
Boiling Pt: 253
Melting Pt: 47
Density: 1.39
MDL Number: MFCD00075057
CAS Number: 50868-72-9
EINECS: 256-816-4

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 42.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR