6-Bromo-m-toluidine ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4507-25G B4507-5G
TCB4507-25GEA 319 CAD
TCB4507-25G TCB4507-5G
6-Bromo-m-toluidine ≥97.0% (by GC, titration analysis)
6-Bromo-m-toluidine
Formula: C₇H₈BrN
Boiling Pt: 130°C
Storage Temperature: Refrigerator
MDL Number: MFCD01806278
CAS Number: 53078-85-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR