(3,5-Dimethoxyphenyl)acetonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3,5-Dimethoxyphenylacetonitrile, 3,5-Dimethoxybenzyl cyanide, 2-(3,5-Dimethoxyphenyl)acetonitrile

D5186-1G D5186-5G
TCD5186-1GEA 82.16 CAD
TCD5186-1G TCD5186-5G
(3,5-Dimethoxyphenyl)acetonitrile ≥98.0% (by GC)
(3,5-Dimethoxyphenyl)acetonitrile
Formula: C₁₀H₁₁NO₂
MW: 177.2
Boiling Pt: 298
Melting Pt: 53
MDL Number: MFCD00016395
CAS Number: 13388-75-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR