4-Fluorobenzoylacetonitrile ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
F0987-1G
F0987-5G
TCF0987-1GEA
75.84
CAD
TCF0987-1G
TCF0987-5G
4-Fluorobenzoylacetonitrile ≥98.0% (by GC)
4-Fluorobenzoylacetonitrile
Formula:
C₉H₆FNO |
MDL Number:
MFCD00662062 CAS Number: 4640-67-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...