1-Naphthylmethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

N1042-25G N1042-5G
76220-368EA 902.18 CAD
76220-368 76220-370
1-Naphthylmethylamine ≥98.0% (by GC, titration analysis)
1-Naphthylmethylamine
Formula: C₁₁H₁₁N
Boiling Pt: 293°C
MDL Number: MFCD00004048
CAS Number: 118-31-0
EINECS: 204-244-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR