N-(6-Bromohexyl)phthalimide ≥98.0% (by GC)

Supplier: TCI America

B4987-25G B4987-5G
TCB4987-25GEA 399.74 CAD
TCB4987-25G TCB4987-5G
N-(6-Bromohexyl)phthalimide ≥98.0% (by GC)
N-(6-Bromohexyl)phthalimide
Formula: C₁₄H₁₆BrNO₂
Boiling Pt: 188°C
MDL Number: MFCD00023098
CAS Number: 24566-79-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR