N-Methylcyclohexylamine ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: N-Cyclohexylmethylamine, Cyclohexyl(methyl)amine

M0202-25ML M0202-500ML
TCM0202-25MLEA 36.34 CAD
TCM0202-25ML TCM0202-500ML
N-Methylcyclohexylamine ≥99.0% (by titrimetric analysis)
N-Methylcyclohexylamine
Formula: C₇H₁₅N
Boiling Pt: 148°C
Flash Pt: 29°C
MDL Number: MFCD00003832
CAS Number: 100-60-7
EINECS: 202-869-3
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR