1-(4-Chlorophenyl)-2-thiourea 98,0 HPLC, N

Supplier: TCI America
Danger

Synonyms: (4-Chlorophenyl)thiourea, 1-(4-Chlorophenyl)thiourea, 4-Chlorophenylthiourea, N-(4-Chlorophenyl)thiourea

C2960-25G C2960-5G
TCC2960-25GEA 864.26 CAD
TCC2960-25G TCC2960-5G
1-(4-Chlorophenyl)-2-thiourea 98,0 HPLC, N
1-(4-Chlorophenyl)-2-thiourea
Formula: (C₆H₄Cl)NC(S)NH₂
MW: 186.66 g/mol
Boiling Pt: 304
MDL Number: MFCD00022168
CAS Number: 3696-23-9
EINECS: 223-022-4
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 169.0 to 173.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR