1-(3-Bromophenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Danger

B4998-25G B4998-5G
TCB4998-25GEA 314.1 CAD
TCB4998-25G TCB4998-5G
1-(3-Bromophenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
1-(3-Bromophenyl)-2-thiourea
Formula: (C₆H₄Br)NHC(S)NH₂
MDL Number: MFCD00060440
CAS Number: 21327-14-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 145.0 to 149.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR