2,4-Dichloro-α-methylbenzyl alcohol ≥98.0% (by GC)

Supplier: TCI America

D4508-25G D4508-5G
TCD4508-25GEA 121.66 CAD
TCD4508-25G TCD4508-5G
2,4-Dichloro-α-methylbenzyl alcohol ≥98.0% (by GC)
2,4-Dichloro-α-methylbenzyl alcohol
Formula: C₈H₈Cl₂O
Boiling Pt: 126°C
Flash Pt: 108°C
MDL Number: MFCD00004511
CAS Number: 1475-13-4
EINECS: 210-019-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR