Gallium(III) acetylacetonate ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T3652-25G T3652-5G
TCT3652-25GEA 396.99 CAD
TCT3652-25G TCT3652-5G
Gallium(III) acetylacetonate ≥97.0% (by titrimetric analysis)
Gallium (III) acetylacetonate
Formula: C₁₅H₂₁GaO₆
Melting Pt: 200°C
MDL Number: MFCD00013492
CAS Number: 14405-43-7
EINECS: 238-377-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystaline
Purity (Chelometric Titration) min. 97.0 %
Melting point 196.0 to 200.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR