1-(2-Nitrophenyl)piperazine ≥98.0% (by GC)

Supplier: TCI America
Warning

N0935-25G N0935-5G
76220-360EA 278.08 CAD
76220-360 76220-362
1-(2-Nitrophenyl)piperazine ≥98.0% (by GC)
1-(2-Nitrophenyl)piperazine
Formula: C₁₀H₁₃N₃O₂
MW: 207.23
Boiling Pt: 126°C
MDL Number: MFCD00040728
CAS Number: 59084-06-9
EINECS: 261-593-1

Order Now

Specification Test Results

Appearance Orange to Red clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR