3'-Chloro-4'-fluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

C3346-25G C3346-5G
TCC3346-25GEA 183.28 CAD
TCC3346-25G TCC3346-5G
3'-Chloro-4'-fluoroacetophenone ≥98.0% (by GC)
3'-Chloro-4'-fluoroacetophenone
Formula: C₈H₆ClFO
MDL Number: MFCD00042203
CAS Number: 2923-66-2

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR