2-Bromo-6-methoxyphenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B5026-1G
B5026-25G
B5026-5G
TCB5026-1GEA
158
CAD
TCB5026-1G
TCB5026-25G
TCB5026-5G
2-Bromo-6-methoxyphenol ≥98.0% (by GC, titration analysis)
2-Bromo-6-methoxyphenol
Formula:
C₇H₇BrO₂ Boiling Pt: 146°C |
MDL Number:
MFCD08146628 CAS Number: 28165-49-3 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 62.0 to 66.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...