Tin(II) 2-ethylhexanoate ≥85.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-Ethylcaproic acid tin (II) salt, Tin bis(2-ethylhexanoate), Stannous 2-ethylhexanoate, 2-Ethylhexanoic acid tin (II) salt

T3149-100G T3149-500G
TCT3149-100GEA 56.88 CAD
TCT3149-100G TCT3149-500G
Tin(II) 2-ethylhexanoate ≥85.0% (by titrimetric analysis)
Tin (II) 2-ethylhexanoate
Formula: C₁₆H₃₀O₄Sn
MW: 405.12 g/mol
MDL Number: MFCD00002676
CAS Number: 301-10-0
EINECS: 206-108-6

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (Iodometric Titration) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR