Pentacene, purified by sublimation

Supplier: TCI America

P2524-100MG P2524-1G
TCP2524-100MGEA 123.4 CAD
TCP2524-100MG TCP2524-1G
Pentacene, purified by sublimation
Pentacene
Formula: C₂₂H₁₄
MDL Number: MFCD00003710
CAS Number: 135-48-8
EINECS: 205-193-7
Merck Index: 14,07107

Order Now

Specification Test Results

Appearance Gray to Dark purple to Black powder to crystal
Total Metals Impurities max. 10.0 ppm
Hole Mobility (mu FET) min. 0.35 cm2/Vs (bare Si/SiO2 substrate)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR