Dimethyltin oxide ≥95.0% (W)

Supplier: TCI America
Danger

D4649-100G D4649-25G
TCD4649-100GEA 513.5 CAD
TCD4649-100G TCD4649-25G
Dimethyltin oxide ≥95.0% (W)
Dimethyltin oxide
Formula: C₂H₆OSn
Melting Pt: 395°C
MDL Number: MFCD00013838
CAS Number: 2273-45-2
EINECS: 218-881-7
UN: 3146
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with ignition residue) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR