2-Cyclohexylethanol ≥98.0% (by GC)

Supplier: TCI America
Warning

C0582-25ML C0582-500ML
TCC0582-25MLEA 45.82 CAD
TCC0582-25ML TCC0582-500ML
2-Cyclohexylethanol ≥98.0% (by GC)
2-Cyclohexylethanol
Formula: C₈H₁₆O
Boiling Pt: 206°C
Flash Pt: 86°C
MDL Number: MFCD00001525
CAS Number: 4442-79-9
EINECS: 224-672-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR