2-Iodobiphenyl ≥98.0% (by GC)
Supplier: TCI America
I0967-25G
I0967-5G
TCI0967-25GEA
453.46
CAD
TCI0967-25G
TCI0967-5G
2-Iodobiphenyl ≥98.0% (by GC)
2-Iodobiphenyl
Formula:
C₁₂H₉I Boiling Pt: 158°C |
MDL Number:
MFCD00039396 CAS Number: 2113-51-1 EINECS: 218-303-3 |
Specification Test Results
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...