3-Methyl-4-nitrobenzyl alcohol ≥98.0% (by GC)

Supplier: TCI America

M2915-1G M2915-5G
TCM2915-1GEA 63.2 CAD
TCM2915-1G TCM2915-5G
3-Methyl-4-nitrobenzyl alcohol ≥98.0% (by GC)
3-Methyl-4-nitrobenzyl alcohol
Formula: C₈H₉NO₃
MDL Number: MFCD00007171
CAS Number: 80866-75-7
EINECS: 279-578-3

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR