4-Methoxy-3-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M3095-25G M3095-5G
TCM3095-25GEA 222.78 CAD
TCM3095-25G TCM3095-5G
4-Methoxy-3-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)
4-Methoxy-3-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₃
MDL Number: MFCD01091014
CAS Number: 213598-09-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 170.0 to 174.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR