4-Nitro-α,α,α-trifluoro-o-toluic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Nitro-2-(trifluoromethyl)benzoic acid

N1037-1G N1037-5G
TCN1037-1GEA 51.19 CAD
TCN1037-1G TCN1037-5G
4-Nitro-α,α,α-trifluoro-o-toluic acid ≥97.0% (by GC, titration analysis)
4-Nitro-α,α,α-trifluoro-o-toluic acid
Formula: C₈H₄F₃NO₄
MW: 235.12
MDL Number: MFCD04039246
CAS Number: 320-37-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 139.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR