4-Methoxy-m-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2728-1G M2728-5G
TCM2728-1GEA 83.74 CAD
TCM2728-1G TCM2728-5G
4-Methoxy-m-toluic acid ≥98.0% (by GC, titration analysis)
4-Methoxy-m-toluic acid
Formula: C₉H₁₀O₃
MDL Number: MFCD00270114
CAS Number: 6880-04-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystaline
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 195.0 to 199.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR