4-Methoxy-o-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2780-1G M2780-25G M2780-5G
TCM2780-1GEA 48.98 CAD
TCM2780-1G TCM2780-25G TCM2780-5G
4-Methoxy-o-toluic acid ≥98.0% (by GC, titration analysis)
4-Methoxy-o-toluic acid
Formula: C₉H₁₀O₃
MDL Number: MFCD00020291
CAS Number: 6245-57-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 178.0 to 182.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR