4-Bromo-3-iodotoluene ≥96.0% (by GC)

Supplier: TCI America
Warning

B5168-1G B5168-5G
TCB5168-1GEA 129.56 CAD
TCB5168-1G TCB5168-5G
4-Bromo-3-iodotoluene ≥96.0% (by GC)
4-Bromo-3-iodotoluene
Formula: C₇H₆BrI
Boiling Pt: 150°C
MDL Number: MFCD07784420
CAS Number: 858841-53-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR